4-(Hydroxymethyl)benzonitril, Sigma-Aldrich®

Lieferant: Merck

SIAL527750-10GEA 587 EUR
SIAL527750-10G
4-(Hydroxymethyl)benzonitril, Sigma-Aldrich®
4-(Hydroxymethyl)benzonitril
Formel: C₈H₇NO
Molecular Weight: 133,15 g/mol
MDL: MFCD00870633
CAS-Nummer: 874-89-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR