3-(Hydroxymethyl)benzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

SIAL09663-10GEA 180 EUR
SIAL09663-10G
3-(Hydroxymethyl)benzonitril, Sigma-Aldrich®
3-(Hydroxymethyl)benzonitril
Formel: C₈H₇NO
Molecular Weight: 133,15 g/mol
MDL: MFCD01321087
CAS-Nummer: 874-97-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR