2-Methoxyisonicotinsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL703680-1GEA 66.6 EUR
SIAL703680-1G
2-Methoxyisonicotinsäure, Sigma-Aldrich®
2-Methoxyisonicotinsäure
Formel: C₇H₇NO₃
Molecular Weight: 153,1400 g/mol
MDL: MFCD04115718
CAS-Nummer: 105596-63-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR