2-Propylthiazol

Lieferant: Thermo Scientific

MAYBSB01111.50EA 0 EUR
MAYBSB01111.50
2-Propylthiazol
2-Propylthiazole
Formel: C₆H₉NS
Molecular Weight: 127,20
MDL: MFCD00053154
CAS-Nummer: 17626-75-4
EINECS: 241-606-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR