2-Chlor-4-(trifluormethyl)benzaldehyd ≥97%

Achtung

H33672.MDEA 54.1 EUR
H33672.MD H33672.03
2-Chlor-4-(trifluormethyl)benzaldehyd ≥97%
2-Chloro-4-(trifluoromethyl)benzaldehyde
Formel: C₈H₄ClF₃O
Molecular Weight: 208,56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04112538
CAS-Nummer: 82096-91-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR