4-Chlor-1H-indazol, Sigma-Aldrich®

Lieferant: Merck

SIAL729922-1GEA 55.5 EUR
SIAL729922-1G
4-Chlor-1H-indazol, Sigma-Aldrich®
4-Chloro-1H-indazole
Formel: C₇H₅ClN₂
Molecular Weight: 152,58 g/mol
MDL: MFCD00211065
CAS-Nummer: 13096-96-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR