4-Chlor-1H-indazol

Lieferant: COMBI-BLOCKS INC.

COBBHI-1348-1GEA 75 EUR
COBBHI-1348-1G COBBHI-1348-5G
4-Chlor-1H-indazol
4-Chloro-1H-indazole
Formel: C₇H₅ClN₂
MDL: MFCD00211065
CAS-Nummer: 13096-96-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR