Diphenylzink, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL481076-1GEA 199 EUR
SIAL481076-1G
Diphenylzink, Sigma-Aldrich®
Diphenylzinc
Formel: C₁₂H₁₀Zn
Molecular Weight: 219,6 g/mol
MDL: MFCD00014071
CAS-Nummer: 1078-58-6
EINECS: 214-082-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR