Diethylzink, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL256781-100GEA 289 EUR
SIAL256781-100G
Diethylzink, Sigma-Aldrich®
Diethylzinc
Formel: C₄H₁₀Zn
Molecular Weight: 123,51 g/mol
MDL: MFCD00009021
CAS-Nummer: 557-20-0
EINECS: 209-161-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR