4-Butylbenzolsulfonamid

Lieferant: Thermo Scientific

MAYBTL00879.1EA 0 EUR
MAYBTL00879.1
4-Butylbenzolsulfonamid
4-Butylbenzolsulfonamid
Formel: C₁₀H₁₅NO₂S
Molecular Weight: 213,29
MDL: MFCD00465202
CAS-Nummer: 1135-00-8
EINECS: 000-000-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR