2-Chlor-6-methylbenzonitril, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL125083-5GEA
105
EUR
SIAL125083-5G
2-Chlor-6-methylbenzonitril, Sigma-Aldrich®
2-Chlor-6-methylbenzonitril
Formel:
C₈H₆ClN Molecular Weight: 151,60 g/mol |
MDL:
MFCD00001782 CAS-Nummer: 6575-09-3 EINECS: 229-499-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...