tert-Butyl-3-brombenzoat

Lieferant: COMBI-BLOCKS INC.

COBBOT-8052-1GEA 36.9 EUR
COBBOT-8052-1G
tert-Butyl-3-brombenzoat
tert-Butyl-3-brombenzoat
Formel: C₁₁H₁₃BrO₂
Molecular Weight: 257,1200 g/mol
MDL: MFCD06657802
CAS-Nummer: 69038-74-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR