Chlorwasserstoff 1.25 mol/l in Ethanol, Supelco®

Lieferant: Merck

Synonyme: Chlorwasserstoff

M-Clarity™ Qualitätsstufe = MQ100

17934-50MLEA 35.7 EUR
17934-50ML 17934-250ML
Chlorwasserstoff 1.25 mol/l in Ethanol, Supelco®
Chlorwasserstoff
Formel: HCl
Molecular Weight: 36,46 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011324
CAS-Nummer: 7647-01-0
EINECS: 231-595-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR