4,5,6,7-Tetrahydro-1H-indazol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL445525-5GEA 73.5 EUR
SIAL445525-5G
4,5,6,7-Tetrahydro-1H-indazol, Sigma-Aldrich®
4,5,6,7-Tetrahydro-1H-indazol
Formel: C₇H₁₀N₂
Molecular Weight: 122,17
MDL: MFCD00014565
CAS-Nummer: 2305-79-5
EINECS: 218-977-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR