2,2-Difluor-2-(2-(trifluormethoxy)phenyl)essigsäure

Lieferant: COMBI-BLOCKS INC.

COBBHC-2556-1GEA 342 EUR
COBBHC-2556-1G
2,2-Difluor-2-(2-(trifluormethoxy)phenyl)essigsäure
2,2-Difluor-2-(2-(trifluormethoxy)phenyl)essigsäure
Formel: C₉H₅F₅O₃
MDL: MFCD11855928
CAS-Nummer: 1133116-03-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR