3-Butoxypropionitril

Lieferant: CHEMPUR

CHMPFL13463.POREA 0 EUR
CHMPFL13463.POR
3-Butoxypropionitril
3-Butoxypropionitril
Formel: C₇H₁₃NO
Molecular Weight: 127,19 g/mol
MDL: MFCD00045643
CAS-Nummer: 6959-71-3
EINECS: 230-153-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR