3,4,6-Trichlorpyridazin

Lieferant: COMBI-BLOCKS INC.

COBBOR-1776-5GEA 0 EUR
COBBOR-1776-5G
3,4,6-Trichlorpyridazin
3,4,6-Trichloropyridazine
Formel: C₄HCl₃N₂
Molecular Weight: 183,4231 g/mol
MDL: MFCD00834957
CAS-Nummer: 6082-66-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR