Isochinolin-3-amin

MAYBBTB10019DAEA 0 EUR
MAYBBTB10019DA MAYBBTB10019CB
Isochinolin-3-amin
Isochinolin-3-amin
Formel: C₉H₈N₂
Molecular Weight: 144,1700 g/mol
MDL: MFCD00102190
CAS-Nummer: 25475-67-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR