Iodmethan-[D] (98% D) stabilisiert, Sigma-Aldrich®
Lieferant: Merck
Stabilisierung:
Stabilised with copper
M-Clarity™ Qualitätsstufe = MQ200
SIAL492825-5GEA
873
EUR
SIAL492825-5G
Iodmethan-[D] (98% D) stabilisiert, Sigma-Aldrich®
Iodmethan-[D]
Formel:
CH₂DI |
MDL:
MFCD00190468 CAS-Nummer: 992-96-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...