[5-(4-Fluorphenyl)-3-isoxazolyl]methanol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL644641-1GEA 92.1 EUR
SIAL644641-1G
[5-(4-Fluorphenyl)-3-isoxazolyl]methanol, Sigma-Aldrich®
[5-(4-Fluorphenyl)-3-isoxazolyl]methanol
Formel: C₁₀H₈FNO₂
MDL: MFCD06798098
CAS-Nummer: 640291-97-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR