5-(2-Methoxyphenyl)-3-(3-nitrophenyl)-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS INC.

COBBHC-6607-5GEA 329 EUR
COBBHC-6607-5G COBBHC-6607-1G
5-(2-Methoxyphenyl)-3-(3-nitrophenyl)-1,2,4-oxadiazol
5-(2-Methoxyphenyl)-3-(3-nitrophenyl)-1,2,4-oxadiazole
Formel: C₁₅H₁₁N₃O₄
MDL: MFCD01152910
CAS-Nummer: 352341-25-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR