5-(3-Chlorphenyl)-3-(3-nitrophenyl)-1,2,4-oxadiazol
Lieferant: COMBI-BLOCKS INC.
COBBHC-6509-5GEA
432
EUR
COBBHC-6509-5G
COBBHC-6509-1G
5-(3-Chlorphenyl)-3-(3-nitrophenyl)-1,2,4-oxadiazol
5-(3-Chlorophenyl)-3-(3-nitrophenyl)-1,2,4-oxadiazole
Formel:
C₁₄H₈ClN₃O₃ |
MDL:
MFCD03719372 CAS-Nummer: 690988-86-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...