5-(2-Chlorphenyl)-3-(4-nitrophenyl)-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS INC.

COBBHC-6506-5GEA 108 EUR
COBBHC-6506-5G COBBHC-6506-1G
5-(2-Chlorphenyl)-3-(4-nitrophenyl)-1,2,4-oxadiazol
5-(2-Chlorophenyl)-3-(4-nitrophenyl)-1,2,4-oxadiazole
Formel: C₁₄H₈ClN₃O₃
MDL: MFCD03885006
CAS-Nummer: 861238-44-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR