3-(4-Chlorphenyl)-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS INC.

COBBHI-1523-5GEA 108 EUR
COBBHI-1523-5G COBBHI-1523-1G
3-(4-Chlorphenyl)-1,2,4-oxadiazol
3-(4-Chlorophenyl)-1,2,4-oxadiazole
Formel: C₈H₅ClN₂O
MDL: MFCD00121590
CAS-Nummer: 10550-15-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR