3-(4-Bromphenyl)-1,2,4-oxadiazol-5(4H)-on

Lieferant: COMBI-BLOCKS INC.

COBBHC-6562-1GEA 210 EUR
COBBHC-6562-1G COBBHC-6562-5G
3-(4-Bromphenyl)-1,2,4-oxadiazol-5(4H)-on
3-(4-Bromphenyl)-1,2,4-oxadiazol-5(4H)-on
Formel: C₈H₅BrN₂O₂
MDL: MFCD11504920
CAS-Nummer: 16672-19-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR