3-(4-Bromphenyl)-5-(3-nitrophenyl)-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS INC.

COBBHC-6589-1GEA 143 EUR
COBBHC-6589-1G COBBHC-6589-5G
3-(4-Bromphenyl)-5-(3-nitrophenyl)-1,2,4-oxadiazol
3-(4-Bromophenyl)-5-(3-nitrophenyl)-1,2,4-oxadiazole
Formel: C₁₄H₈BrN₃O₃
MDL: MFCD02921806
CAS-Nummer: 429624-16-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR