3-(4-Bromphenyl)-5-(2-chlorphenyl)-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS INC.

COBBHC-6595-1GEA 143 EUR
COBBHC-6595-1G COBBHC-6595-5G
3-(4-Bromphenyl)-5-(2-chlorphenyl)-1,2,4-oxadiazol
3-(4-Bromophenyl)-5-(2-chlorophenyl)-1,2,4-oxadiazole
Formel: C₁₄H₈BrClN₂O
MDL: MFCD00697977
CAS-Nummer: 404900-69-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR