3-(2-Bromphenyl)-5-(3-chlorphenyl)-1,2,4-oxadiazol
Lieferant: COMBI-BLOCKS INC.
COBBHC-6283-5GEA
409
EUR
COBBHC-6283-5G
COBBHC-6283-1G
3-(2-Bromphenyl)-5-(3-chlorphenyl)-1,2,4-oxadiazol
3-(2-Bromophenyl)-5-(3-chlorophenyl)-1,2,4-oxadiazole
Formel:
C₁₄H₈BrClN₂O |
MDL:
MFCD10699652 CAS-Nummer: 1033201-83-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...