5-Cyclohexyl-3-(3-nitrophenyl)-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS INC.

COBBHC-6531-1GEA 72 EUR
COBBHC-6531-1G COBBHC-6531-5G
5-Cyclohexyl-3-(3-nitrophenyl)-1,2,4-oxadiazol
5-Cyclohexyl-3-(3-nitrophenyl)-1,2,4-oxadiazole
Formel: C₁₄H₁₅N₃O₃
MDL: MFCD04208216
CAS-Nummer: 442571-86-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR