5-(Chlormethyl)-3-(4-methylphenyl)-1,2,4-oxadiazol
Lieferant: COMBI-BLOCKS INC.
COBBHC-6504-5GEA
262
EUR
COBBHC-6504-5G
COBBHC-6504-1G
5-(Chlormethyl)-3-(4-methylphenyl)-1,2,4-oxadiazol
5-(Chloromethyl)-3-(4-methylphenyl)-1,2,4-oxadiazole
Formel:
C₁₀H₉ClN₂O |
MDL:
MFCD06093407 CAS-Nummer: 50737-29-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...