5-(Prop-1-en-2-yl)-3-(4-methylphenyl)-1,2,4-oxadiazol
Lieferant: COMBI-BLOCKS INC.
COBBHC-6519-5GEA
432
EUR
COBBHC-6519-5G
COBBHC-6519-1G
5-(Prop-1-en-2-yl)-3-(4-methylphenyl)-1,2,4-oxadiazol
5-(Prop-1-en-2-yl)-3-(4-methylphenyl)-1,2,4-oxadiazol
Formel:
C₁₂H₁₂N₂O |
MDL:
MFCD10699661 CAS-Nummer: 1033201-98-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...