6-(4-Fluorphenyl)-3-pyridinboronsäurepinakolester 95%

Lieferant: Thermo Scientific
Achtung

H27168.MDEA 152 EUR
H27168.MD
6-(4-Fluorphenyl)-3-pyridinboronsäurepinakolester 95%
6-(4-Fluorphenyl)-3-pyridinboronsäurepinakolester
Formel: C₁₇H₁₉BFNO₂
Molecular Weight: 299,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09037494
CAS-Nummer: 1073354-81-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR