2-(4-Methoxyphenyl)isonicotinsäure

Lieferant: COMBI-BLOCKS INC.

COBBYA-6431-1GEA 630 EUR
COBBYA-6431-1G
2-(4-Methoxyphenyl)isonicotinsäure
2-(4-Methoxyphenyl)isonicotinsäure
Formel: C₁₃H₁₁NO₃
MDL: MFCD09909443
CAS-Nummer: 935861-30-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR