2-Acetylamino-5-brom-6-methylpyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL643475-1GEA 88 EUR
SIAL643475-1G
2-Acetylamino-5-brom-6-methylpyridin, Sigma-Aldrich®
2-Acetylamino-5-brom-6-methylpyridin
Formel: C₈H₉BrN₂O
Molecular Weight: 229,0700 g/mol
MDL: MFCD03909755
CAS-Nummer: 142404-84-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR