6-Hydroxy-5-nitronicotinonitril

Lieferant: COMBI-BLOCKS INC.

COBBPY-7821-1GEA 1180 EUR
COBBPY-7821-1G
6-Hydroxy-5-nitronicotinonitril
6-Hydroxy-5-nitronicotinonitril
Formel: C₆H₃N₃O₃
MDL: MFCD11504985
CAS-Nummer: 320405-84-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR