2-Brom-N-cyclopropylisonicotinamid

Lieferant: COMBI-BLOCKS INC.

COBBPY-7229-5GEA 432 EUR
COBBPY-7229-5G COBBPY-7229-25G COBBPY-7229-1G
2-Brom-N-cyclopropylisonicotinamid
2-Brom-N-cyclopropylisonicotinamid
Formel: C₉H₉BrN₂O
MDL: MFCD09878430
CAS-Nummer: 951885-74-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR