2-Brom-N-ethylisonicotinamid
Lieferant: COMBI-BLOCKS INC.
COBBPY-7228-1GEA
117
EUR
COBBPY-7228-1G
COBBPY-7228-5G
COBBPY-7228-25G
2-Brom-N-ethylisonicotinamid
2-Brom-N-ethylisonicotinamid
Formel:
C₈H₉BrN₂O |
MDL:
MFCD09878429 CAS-Nummer: 951885-78-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...