2-(Cyclopentylthio)-5-pyridinboronsäure
Lieferant: COMBI-BLOCKS INC.
COBBBB-6241-250MEA
0
EUR
COBBBB-6241-250M
COBBBB-6241-1G
2-(Cyclopentylthio)-5-pyridinboronsäure
2-(Cyclopentylthio)-5-pyridinboronsäure
Formel:
C₁₀H₁₄BNO₂S |
MDL:
MFCD15143474 CAS-Nummer: 1218790-70-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...