5-Amino-1-chlorisochinolin

Lieferant: COMBI-BLOCKS INC.

COBBHC-6029-1GEA 571 EUR
COBBHC-6029-1G
5-Amino-1-chlorisochinolin
5-Amino-1-chloroisoquinoline
Formel: C₉H₇ClN₂
Molecular Weight: 178,6183 g/mol
MDL: MFCD09258640
CAS-Nummer: 374554-54-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR