6-Brom-7-chlor-8-methylchinolin
Lieferant: COMBI-BLOCKS INC.
COBBHC-6958-1GEA
143
EUR
COBBHC-6958-1G
COBBHC-6958-5G
6-Brom-7-chlor-8-methylchinolin
6-Bromo-7-chloro-8-methylquinoline
Formel:
C₁₀H₇BrClN |
MDL:
MFCD10699677 CAS-Nummer: 1033202-16-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...