2-Methyl-7-chinolinboronsäurepinakolester 96%

Achtung

H54984.MDEA 114.4 EUR
H54984.MD
2-Methyl-7-chinolinboronsäurepinakolester 96%
2-Methyl-7-chinolinboronsäurepinakolester
Formel: C₁₆H₂₀BNO₂
Molecular Weight: 269,15 g/mol
Schmelzpunkt: 100…102 °C
MDL: MFCD11846348
CAS-Nummer: 939052-79-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR