2-Brom-8-chlorchinolin
Lieferant: COMBI-BLOCKS INC.
COBBPY-1364-1GEA
368
EUR
COBBPY-1364-1G
COBBPY-1364-5G
COBBPY-1364-250M
2-Brom-8-chlorchinolin
2-Bromo-8-chloroquinoline
Formel:
C₉H₅BrClN |
MDL:
MFCD13689182 CAS-Nummer: 891842-52-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...