2-Benzyloxy-4-fluorphenylboronsäure ≥98%

Lieferant: Thermo Scientific
Achtung

H53002.06EA 74 EUR
H53002.06 H53002.14 H53002.03
2-Benzyloxy-4-fluorphenylboronsäure ≥98%
2-Benzyloxy-4-fluorphenylboronsäure
Formel: C₁₃H₁₂BFO₃
Molecular Weight: 246,04
Schmelzpunkt: 110…116 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03788402
CAS-Nummer: 848779-87-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR