(4-Benzamidophenyl)boronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-2934-1GEA 275 EUR
COBBBB-2934-1G COBBBB-2934-5G
(4-Benzamidophenyl)boronsäure
(4-Benzamidophenyl)boronsäure
Formel: C₁₃H₁₂BNO₃
Molecular Weight: 241,05 g/mol
MDL: MFCD08235089
CAS-Nummer: 397843-80-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR