4-Cyclopentylphenylboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-9034-1GEA 341 EUR
COBBBB-9034-1G COBBBB-9034-5G
4-Cyclopentylphenylboronsäure
4-Cyclopentylphenylboronsäure
Formel: C₁₁H₁₅BO₂
MDL: MFCD04039014
CAS-Nummer: 945837-57-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR