1-Benzyloxy-3-methyl-2-nitrobenzol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL510521-5GEA 104 EUR
SIAL510521-5G
1-Benzyloxy-3-methyl-2-nitrobenzol, Sigma-Aldrich®
1-Benzyloxy-3-methyl-2-nitrobenzol
Formel: C₁₄H₁₃NO₃
Molecular Weight: 243,2579 g/mol
MDL: MFCD00272556
CAS-Nummer: 61535-21-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR