4-(2-Aminoethyl)benzolsulfonamid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL275247-25GEA 56.6 EUR
SIAL275247-25G
4-(2-Aminoethyl)benzolsulfonamid, Sigma-Aldrich®
4-(2-Aminoethyl)benzolsulfonamid
Formel: C₈H₁₂N₂O₂S
Molecular Weight: 200,26
MDL: MFCD00010301
CAS-Nummer: 35303-76-5
EINECS: 252-501-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR