4-(2-Aminoethyl)benzolsulfonamid

Lieferant: Molekula

MOLE16359200-100GEA 0 EUR
MOLE16359200-100G
4-(2-Aminoethyl)benzolsulfonamid
4-(2-Aminoethyl)benzolsulfonamid
Formel: C₈H₁₂N₂O₂S
Molecular Weight: 200,26 g/mol
MDL: MFCD00010301
CAS-Nummer: 35303-76-5
EINECS: 252-501-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR