2,6-Dimethoxybenzamidin Hydrochlorid

Lieferant: CHEMPUR

CHMP12-0044.1EA 0 EUR
CHMP12-0044.1 CHMP12-0044.5
2,6-Dimethoxybenzamidin Hydrochlorid
2,6-Dimethoxybenzamidin Hydrochlorid
Formel: C₉H₁₃ClN₂O₂
MDL: MFCD04114428
CAS-Nummer: 1171047-31-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR