2,4-Dimethoxybenzamidin Hydrochlorid

Lieferant: CHEMPUR

CHMP12-0041.1EA 0 EUR
CHMP12-0041.1 CHMP12-0041.5
2,4-Dimethoxybenzamidin Hydrochlorid
2,4-Dimethoxybenzamidin Hydrochlorid
Formel: C₉H₁₃ClN₂O₂
MDL: MFCD04114426
CAS-Nummer: 131947-81-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR